Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 2 - 3 | 395 | 91 | |
NM_019671.3 | NP_062645.2 | 4 - 5 | 516 | 91 | ||
GenBank mRNA | AF094520.1 | - | 4 - 5 | 412 | 91 | |
AJ010045.1 | - | 2 - 3 | 358 | 91 | ||
AK088632.1 | - | 2 - 3 | 359 | 91 | ||
AK139921.1 | - | 2 - 3 | 359 | 91 | ||
BC004699.1 | - | 2 - 3 | 369 | 3499258 | 91 | |
CN702942.1 | - | 2 - 3 | 256 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
