Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 6 - 7 | 793 | 93 | |
NM_019671.3 | NP_062645.2 | 8 - 9 | 914 | 93 | ||
GenBank mRNA | AF094520.1 | - | 8 - 9 | 810 | 93 | |
AJ010045.1 | - | 6 - 7 | 756 | 93 | ||
AK082128.1 | - | 6 - 7 | 790 | 93 | ||
AK088632.1 | - | 6 - 7 | 757 | 93 | ||
AK139921.1 | - | 6 - 7 | 757 | 93 | ||
BC004699.1 | - | 6 - 7 | 767 | 3499258 | 93 | |
CA980605.1 | - | 2 - 3 | 96 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
