Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 7 - 8 | 1057 | 90 | |
NM_019671.3 | NP_062645.2 | 9 - 10 | 1178 | 90 | ||
GenBank mRNA | AF094520.1 | - | 9 - 10 | 1074 | 90 | |
AJ010045.1 | - | 7 - 8 | 1020 | 90 | ||
AK082128.1 | - | 7 - 8 | 1054 | 90 | ||
AK088632.1 | - | 7 - 8 | 1021 | 90 | ||
AK139921.1 | - | 7 - 8 | 1021 | 90 | ||
BC004699.1 | - | 7 - 8 | 1031 | 3499258 | 90 | |
CA980605.1 | - | 3 - 4 | 360 | 90 | ||
CN697204.1 | - | 1 - 2 | 177 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
