Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025779.1 | NP_001020950.1 | 4 - 5 | 767 | 86 | |
NM_011799.2 | NP_035929.1 | 4 - 5 | 843 | 86 | ||
GenBank mRNA | AJ009559.1 | - | 4 - 5 | 860 | 86 | |
AJ223087.1 | - | 4 - 5 | 845 | 86 | ||
AK012192.1 | - | 4 - 5 | 797 | 86 | ||
AK075975.1 | - | 4 - 5 | 796 | 86 | ||
AK085020.1 | - | 4 - 5 | 797 | 86 | ||
AK166003.1 | - | 4 - 5 | 848 | 86 | ||
BC052434.1 | - | 4 - 5 | 767 | 6837113 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
