Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 3 - 4 | 553 | 120 | |
GenBank mRNA | AK142280.1 | - | 3 - 4 | 545 | 120 | |
AK150089.1 | - | 3 - 4 | 545 | 120 | ||
AK157252.1 | - | 3 - 4 | 515 | 120 | ||
AK160880.1 | - | 3 - 4 | 545 | 120 | ||
AK160890.1 | - | 3 - 4 | 531 | 120 | ||
AK160973.1 | - | 3 - 4 | 531 | 120 | ||
AK167582.1 | - | 3 - 4 | 541 | 120 | ||
AK191948.1 | - | 2 - 3 | 115 | 120 | ||
AK203724.1 | - | 2 - 3 | 115 | 120 | ||
AK210573.1 | - | 2 - 3 | 120 | |||
AK218197.1 | - | 2 - 3 | 115 | 120 | ||
BC027747.1 | - | 3 - 4 | 4459657 | 120 | ||
BC068193.1 | - | 3 - 4 | 512 | 6813997 | 120 | |
X92411.1 | - | 3 - 4 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
