Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 4 - 5 | 814 | 104 | |
GenBank mRNA | AK142280.1 | - | 4 - 5 | 806 | 104 | |
AK150089.1 | - | 4 - 5 | 806 | 104 | ||
AK157252.1 | - | 4 - 5 | 776 | 104 | ||
AK160880.1 | - | 4 - 5 | 806 | 104 | ||
AK160890.1 | - | 4 - 5 | 792 | 104 | ||
AK160973.1 | - | 4 - 5 | 792 | 104 | ||
AK167582.1 | - | 4 - 5 | 802 | 104 | ||
BC027747.1 | - | 4 - 5 | 803 | 4459657 | 104 | |
BC068193.1 | - | 4 - 5 | 773 | 6813997 | 104 | |
X92411.1 | - | 4 - 5 | 494 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
