Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 6 - 7 | 992 | 77 | |
GenBank mRNA | AK142280.1 | - | 6 - 7 | 984 | 77 | |
AK150089.1 | - | 6 - 7 | 984 | 77 | ||
AK157252.1 | - | 6 - 7 | 954 | 77 | ||
AK160880.1 | - | 6 - 7 | 984 | 77 | ||
AK160890.1 | - | 6 - 7 | 970 | 77 | ||
AK160973.1 | - | 6 - 7 | 970 | 77 | ||
AK167582.1 | - | 6 - 7 | 980 | 77 | ||
AK198214.1 | - | 2 - 3 | 150 | 77 | ||
AK218725.1 | - | 2 - 3 | 156 | 77 | ||
BC027747.1 | - | 6 - 7 | 981 | 4459657 | 77 | |
BC068193.1 | - | 6 - 7 | 951 | 6813997 | 77 | |
X92411.1 | - | 6 - 7 | 672 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
