Assay Details
Gene Symbol: Btnl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030746.1 | NP_109671.1 | 5 - 6 | 941 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962532 | NM_030746.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69865 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962532 | NM_030746.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31154051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154069 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13412889 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13124301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154082 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13229189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13124292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154077 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154090 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13231524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31154089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13124295 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450176 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450173 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450172 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450175 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450178 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450179 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450177 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450174 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450167 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700017841.mm.1 | TC1700001909.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017841.mm.1 | NM_030746.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017841.mm.1 | NM_030746.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017843.mm.1 | TC1700001909.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017843.mm.1 | NM_030746.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017843.mm.1 | NM_030746.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017844.mm.1 | TC1700001909.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017844.mm.1 | NM_030746.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017844.mm.1 | NM_030746.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017842.mm.1 | TC1700001909.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700017842.mm.1 | NM_030746.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700017842.mm.1 | NM_030746.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 883027 | - | - |
