Assay Details
Gene Symbol: Uchl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033607.1 | NP_291085.1 | 1 | 354 | 65 | |
GenBank mRNA | AB035420.1 | - | 1 | 354 | 65 | |
BC139440.1 | - | 1 | 345 | 8862462 | 65 | |
BC139441.1 | - | 1 | 345 | 8862463 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172455 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | AB035420.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | BC139440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | BC139441.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | AB035420.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | BC139440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580874 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26580868 | - | - |
Whole Mouse Genome G4122A | A_52_P315631 | NM_033607.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P315631 | AB035420.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P315631 | BC139440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P315631 | BC139441.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | NM_033607.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | AB035420.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | BC139440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P395921 | BC139441.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07457106 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07887844 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07887835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26580887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580897 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07457107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07501392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580846 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07887835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07626080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07887832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07621007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07696024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07621012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07621015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07696025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26580878 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10586183 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586181 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586182 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421766_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10586180 | - | - |
Mouse Genome 430 2.0 Array | 1421766_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1421766_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93841_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93841_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900006099.mm.1 | TC0900000818.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900006099.mm.1 | NM_033607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900006099.mm.1 | NM_033607.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900006098.mm.1 | TC0900000818.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900006098.mm.1 | NM_033607.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900006098.mm.1 | NM_033607.1 | Refseq |
Mouse Expression Set 430 | 1421766_at | - | - |
Mouse Expression Set 430 | 1449855_s_at | - | - |
Mouse Expression Set 430 | 1449855_PM_s_at | - | - |
Mouse Expression Set 430 | 1421766_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421766_at | - | - |
Mouse Genome 430A 2.0 Array | 1449855_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1449855_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1421766_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421766_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421766_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 413970 | - | - |
ABI Mouse Genome Survey Microarray | 417091 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1740181 | - | - |
MouseWG-6 v2 Expression BeadChip | 6220110 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1740181 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6220110 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001740181 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006220110 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2485802 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235288 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2494496 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1235288 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2485802 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2494496 | - | - |
mouse-6 v1.1 (Illumina) | scl093841.1_30-S | - | - |
mouse-6 v1.1 (Illumina) | scl0093841.1_114-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6220110 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1740181 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl093841.1_30-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093841.1_114-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740181 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220110 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1235288 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1235288 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2781878 | - | - |
