Assay Details
Gene Symbol: Gm12942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099319.2 | NP_001092789.1 | 2 - 3 | 330 | 114 | |
GenBank mRNA | AK156456.1 | - | 2 - 3 | 312 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019909 | AK160613.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019909 | NM_001099319.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019909 | AK156456.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1180 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963156 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019909 | AK160613.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019909 | NM_001099319.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019909 | AK156456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70216 | AK160613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70216 | NM_001099319.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70216 | AK156456.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23283621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03375190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03668565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03831504 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03208990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03414316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03668560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283662 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03831503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03337948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03933372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03783284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03831516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03337949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03307479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03414315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03809808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03414313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03831512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04044016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03307483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283705 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03208979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283650 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03208984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23283632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10508250 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508252 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10508251 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434193_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10508249 | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100039968_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100039968_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400011355.mm.1 | TC0400001482.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011355.mm.1 | NM_001099319.2 | Refseq |
Mouse Expression Set 430 | 1434193_PM_at | - | - |
Mouse Expression Set 430 | 1434193_at | - | - |
