Assay Details
Gene Symbol: Sec14l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127725.1 | NP_001121197.1 | 10 - 11 | 1196 | 87 | |
GenBank mRNA | BC157907.1 | - | 10 - 11 | 1262 | 9088109 | 87 |
BC157990.1 | - | 10 - 11 | 1196 | 9007304 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
