Assay Details
Gene Symbol: Pnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123371.2 | NP_001116843.1 | 6 | 946 | 121 | |
GenBank mRNA | AK008143.1 | - | 6 | 950 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976898 | AK008143.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976898 | NM_001123371.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16185 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524261 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109371 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976898 | AK008143.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976898 | NM_001123371.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29599388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11501852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11179507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29599315 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10414531 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414528 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414529 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414530 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10414527 | - | - |
Mouse Genome 430 2.0 Array | 1453299_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1453299_a_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400004510.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004510.mm.1 | NM_001123371.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400004509.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004509.mm.1 | NM_001123371.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400004506.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004506.mm.1 | NM_001123371.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400004511.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004511.mm.1 | NM_001123371.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400004508.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004508.mm.1 | NM_001123371.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400004507.mm.1 | TC1400000575.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400004507.mm.1 | NM_001123371.2 | Refseq |
Mouse Expression Set 430 | 1453299_a_at | - | - |
Mouse Expression Set 430 | 1453299_PM_a_at | - | - |
Murine Genome U74v2 Set | 93290_at | - | - |
Murine Genome U74v2 Set | 93290_PM_at | - | - |
Murine Genome U74v2 Set | 165699_PM_r_at | - | - |
Murine Genome U74v2 Set | 165699_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1453299_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1453299_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102570402 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102570402 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570402 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2549473 | - | - |
