Assay Details
Gene Symbol: ATP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF093677.1 | - | 1 | 591 | 110 | |
NC_005089_ATP6.0 | - | 452 | 110 |
Gene Symbol: Gm10925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC069931.1 | - | 2 | 547 | 3489785 | 110 |

More Information
Additional Information:
NC_005089 (strand:+; 7927..8607); ATP synthase F0 subunit 6
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
