Assay Details
Gene Symbol: Btnl5-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_004051.1 | 4 - 5 | 1103 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | butyrophilin-like 4 | Btnl4 |
Btn3a3, EG632126, NG11 |
-15.1kb | Sense | Chr. 17 - 34468410 - 34477428 on NCBI Build GRCm38 |
Downstream | butyrophilin-like 6 | Btnl6 |
EG624681, Gm6519, NG13 |
14.6kb | Sense | Chr. 17 - 34507935 - 34517352 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
