Assay Details
Gene Symbol: 3110070M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027974.1 | 1 | 954 | 69 | ||
GenBank mRNA | AK005613.1 | - | 1 | 946 | 69 | |
AK014250.1 | - | 1 | 954 | 69 | ||
BC050963.1 | - | 1 | 954 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 7120 | Gm7120 | EG633640 | Anti-sense | Chr. 13 - 119488039 - 119610459 on NCBI Build GRCm38 | |
Downstream | chemokine (C-C motif) ligand 28 | Ccl28 |
CCK1, MEC, Scya28 |
122.2kb | Anti-sense | Chr. 13 - 119610551 - 119654359 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
