Assay Details
Gene Symbol: 1700021N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045800.1 | 1 - 2 | 55 | |||
GenBank mRNA | AK006224.1 | - | 1 - 2 | 184 | 55 | |
BY706310.2 | - | 1 - 2 | 187 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | platelet-activating factor acetylhydrolase 2 | Pafah2 |
2310074E22Rik, AI507170 |
-22.3kb | Anti-sense | Chr. 4 - 134396320 - 134427412 on NCBI Build GRCm38 |
Downstream | stathmin 1 | Stmn1 |
19k, Lag, Lap18, Op18, P18, P19, Pig, Pp17, Pp18, Pp19, Pr22, Smn |
18.2kb | Anti-sense | Chr. 4 - 134468320 - 134473843 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
