Assay Details
Gene Symbol: 1700037C06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006618.1 | - | 1 | 745 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 13001 | Gm13001 | OTTMUSG00000009820 | Sense | Chr. 4 - 137116705 - 137118255 on NCBI Build GRCm38 | |
Downstream | predicted gene 13003 | Gm13003 | OTTMUSG00000009822 | 41.5kb | Sense | Chr. 4 - 137159082 - 137165066 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
