Assay Details
Gene Symbol: 1700061A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006847.1 | - | 1 | 343 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MUS81 endonuclease homolog (yeast) | Mus81 |
1200008A18Rik, AI182501, AW045863 |
-335.0 | Sense | Chr. 19 - 5482836 - 5488409 on NCBI Build GRCm38 |
Downstream |
cofilin 1, non-muscle |
Cfl1 |
AA959946, Cof, n-cofilin |
1.3kb | Anti-sense | Chr. 19 - 5490455 - 5494031 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
