Assay Details
Gene Symbol: 2010106G04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008322.1 | - | 1 | 148 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
TBC1 domain family, member 22B |
Tbc1d22b | BC045600 | -7.6kb | Anti-sense | Chr. 17 - 29549788 - 29606894 on NCBI Build GRCm38 |
Downstream | ring finger protein 8 | Rnf8 |
3830404E21Rik, AIP37 |
62.0 | Anti-sense | Chr. 17 - 29614831 - 29641659 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
