Assay Details
Gene Symbol: C030009O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021058.1 | - | 1 | 453 | 86 | |
AK045829.1 | - | 1 | 877 | 86 | ||
AK140124.1 | - | 1 | 927 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 38562 |
Gm38562 | -445.1kb | Anti-sense | Chr. 5 - 71680915 - 71701733 on NCBI Build GRCm38 | |
Downstream | COMM domain containing 8 | Commd8 |
C77770, D5Buc26e |
9.1kb | Anti-sense | Chr. 5 - 72156576 - 72168183 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
