Assay Details
Gene Symbol: 2010310C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028457.1 | - | 53 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Eph receptor A1 | Epha1 |
5730453L17Rik, AL033318, Eph, Esk |
-4.6kb | Anti-sense | Chr. 6 - 42358487 - 42373310 on NCBI Build GRCm38 |
Downstream |
taste receptor, type 2, member 143 |
Tas2r143 |
Gm1404, Tas2r43, mt2r36 |
21.2kb | Sense | Chr. 6 - 42400238 - 42401119 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
