Assay Details
Gene Symbol: E230016M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040278.1 | 6 - 7 | 1006 | 142 | ||
GenBank mRNA | AK029473.1 | - | 6 - 7 | 1008 | 142 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth arrest and DNA-damage-inducible 45 alpha | Gadd45a |
AA545191, Ddit1, GADD45 |
-40.8kb | Anti-sense | Chr. 6 - 67035096 - 67037407 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA A430010J10 gene | A430010J10Rik | 77.1kb | Sense | Chr. 6 - 67157801 - 67169010 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172806 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352299 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885254 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188650 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472995 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859245 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469741 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71771 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554848 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185512 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301971 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357325 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396208 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536579 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91318 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152395 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106881 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AV252933.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AI627090.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AK029473.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AV252933.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AI627090.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P526513 | AK029473.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05928591 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05928572 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05928626 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106881 | - | - |
Whole Mouse Genome G4122A | A_52_P526513 | AV252933.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P526513 | AI627090.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P526513 | AK029473.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P526513 | AV252933.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P526513 | AI627090.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P526513 | AK029473.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P181025 | AK054076.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132801 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05534868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05073072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05440052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05076480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05477121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05440050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05166421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05073075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05534870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05440053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05076492 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05367091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519607 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05625928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05076490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05625926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05073082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24519770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05440056 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1456794_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444637_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1444637_at | - | - |
Mouse Genome 430 2.0 Array | 1456794_at | - | - |
Mouse Genome 430 2.0 Array | 1456794_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1444637_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320172_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320172_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600005850.mm.1 | TC0600000780.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600005850.mm.1 | NR_040278.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005850.mm.1 | NR_040278.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005848.mm.1 | TC0600000780.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600005848.mm.1 | NR_040278.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005848.mm.1 | NR_040278.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005849.mm.1 | TC0600000780.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600005849.mm.1 | NR_040278.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005849.mm.1 | NR_040278.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005851.mm.1 | TC0600000780.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600005851.mm.1 | NR_040278.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600005851.mm.1 | NR_040278.1 | Refseq |
Mouse Expression Set 430 | 1456794_at | - | - |
Mouse Expression Set 430 | 1444637_at | - | - |
Mouse Expression Set 430 | 1444637_PM_at | - | - |
Mouse Expression Set 430 | 1456794_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444637_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444637_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456794_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456794_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 385793 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104010452 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1226631 | - | - |
