Assay Details
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031001.1 | - | 1 | 4250 | 78 | |
AK164078.1 | - | 1 | 3931 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6541 | Mir6541 |
Gm27311, mmu-mir-6541 |
-91.1kb | Sense | Chr. 14 - 62865542 - 62865651 on NCBI Build GRCm38 |
Downstream | defensin beta 48 | Defb48 |
Defb42, EG432867 |
16.4kb | Anti-sense | Chr. 14 - 62977524 - 62984510 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
