Assay Details
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031725.1 | - | 1 | 847 | 89 | |
AK047638.1 | - | 1 | 4018 | 89 | ||
AK081408.1 | - | 1 | 809 | 89 | ||
BC060127.1 | - | 1 | 3555 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tektin 2 | Tekt2 |
AU014974, tektin-t |
-8.8kb | Sense | Chr. 4 - 126322121 - 126325199 on NCBI Build GRCm38 |
Downstream | argonaute RISC catalytic subunit 1 | Ago1 | Eif2c1 | 99.6kb | Sense | Chr. 4 - 126435012 - 126468709 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
