Assay Details
Gene Symbol: 5033428I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034688.1 | - | 2 - 3 | 261 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heart and neural crest derivatives expressed transcript 2 | Hand2 |
AI225906, AI661148, Ehand2, Hed, Th2, Thing2, bHLHa26, dHAND |
-7.9kb | Sense | Chr. 8 - 57320983 - 57324517 on NCBI Build GRCm38 |
Downstream | scrapie responsive gene 1 | Scrg1 | AW124307 | 122.3kb | Sense | Chr. 8 - 57455923 - 57477585 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
