Assay Details
Gene Symbol: 5830428M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038060.1 | 4 - 5 | 406 | 67 | ||
NR_038061.1 | 4 - 5 | 400 | 67 | |||
GenBank mRNA | AI060956.1 | - | 4 - 5 | 381 | 67 | |
AK039829.1 | - | 4 - 5 | 403 | 67 | ||
DV046824.1 | - | 4 - 5 | 386 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ADP-ribosylation factor 6 | Arf6 |
AI788669, AW496366 |
-88.8kb | Anti-sense | Chr. 12 - 69372150 - 69375980 on NCBI Build GRCm38 |
Downstream | valosin containing protein lysine (K) methyltransferase | Vcpkmt |
Gm71, Mettl21d, Mettl221d, VCP-KMT |
108.4kb | Sense | Chr. 12 - 69576730 - 69583109 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2283310 | NR_038061.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2283310 | AK039829.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2283310 | NR_038060.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444193 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137355 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2283310 | NR_038061.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2283310 | AK039829.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2283310 | NR_038060.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137355 | - | - |
Whole Mouse Genome G4122A | A_52_P105020 | NR_038061.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P105020 | AK039829.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P105020 | NR_038060.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10352654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514755 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10006941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10120164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10298439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10043213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514774 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10120163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514738 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514771 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09833518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10080175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09931130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514719 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514734 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10006942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10186547 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514777 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09904803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514680 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10078369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09826775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10080169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10080170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28514713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10164735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10352645 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1440266_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440266_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440266_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200012701.mm.1 | TC1200001866.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200012701.mm.1 | NR_038060.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200012701.mm.1 | NR_038060.1 | Refseq |
Mouse Expression Set 430 | 1440266_at | - | - |
Mouse Expression Set 430 | 1440266_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440266_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440266_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 340591 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 100510333 | - | - |
