Assay Details
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK048719.1 | - | 1 - 2 | 107 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | latent transforming growth factor beta binding protein 4 | Ltbp4 | 2310046A13Rik | -14.6kb | Anti-sense | Chr. 7 - 27305140 - 27339709 on NCBI Build GRCm38 |
Downstream |
spectrin beta, non-erythrocytic 4 |
Sptbn4 |
1700022P15Rik, 5830426A08Rik, ROSA62, SpbIV, Spnb4, dyn, lnd, nmf261, nmf379, qv |
150.0 | Anti-sense | Chr. 7 - 27356383 - 27447704 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
