Assay Details
Gene Symbol: 4930553C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016100.1 | - | 2 - 3 | 615 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 12130 | Gm12130 | OTTMUSG00000005424 | -787.7kb | Sense | Chr. 11 - 38491548 - 38520045 on NCBI Build GRCm38 |
Downstream | predicted gene 12132 | Gm12132 | OTTMUSG00000005426 | 620.7kb | Anti-sense | Chr. 11 - 40008194 - 40337842 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441034 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654255 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790529 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61965 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431937 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958803 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341608 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536766 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229504 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675465 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771972 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 276463 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374945 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478589 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728283 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143588 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83343 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9697 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137805 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07313098 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07313064 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07313037 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02304324 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07312967 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07312967 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07313037 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07313098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09210948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730187 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09428223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09454021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09493643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729889 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730215 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09383533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09305445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730258 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09675679 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09036424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09328120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09243655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09206635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09248295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09210951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09690662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09534853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27729737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27730292 | - | - |
