Assay Details
Gene Symbol: 5830426K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017949.1 | - | 1 | 255 | 95 | |
AK041807.1 | - | 1 | 1297 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase 1E (PP2C domain containing) | Ppm1e |
AW049266, B930008A12Rik, POPX1, PP2CH, mKIAA1072 |
Sense | Chr. 11 - 87226906 - 87358994 on NCBI Build GRCm38 | |
Downstream | RAD51 homolog C | Rad51c |
R51H3, Rad51l2 |
34.8kb | Sense | Chr. 11 - 87377607 - 87404954 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
