Assay Details
Gene Symbol: 4930502C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076703.1 | - | 683 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
C-type lectin domain family 2, member L |
Clec2l |
AI414054, AW122724, Gm1064 |
-93.1kb | Sense | Chr. 6 - 38663069 - 38680865 on NCBI Build GRCm38 |
Downstream |
thromboxane A synthase 1, platelet |
Tbxas1 |
CYP5, CYP5A1, THAS, TS, TXAS, TXS |
142.4kb | Sense | Chr. 6 - 38918467 - 39084590 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
