Assay Details
Gene Symbol: Junos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076859.1 | - | 4 - 5 | 509 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | jun proto-oncogene | Jun |
AP-1, Junc, c-jun |
-109.0kb | Anti-sense | Chr. 4 - 95049036 - 95052222 on NCBI Build GRCm38 |
Downstream | predicted gene 12708 | Gm12708 | OTTMUSG00000008048 | 27.2kb | Sense | Chr. 4 - 95193274 - 95209724 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
