Assay Details
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078829.1 | - | 1 | 51 | 90 | |
AK088511.1 | - | 1 | 2291 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930578N18 gene | 4930578N18Rik | -131.3kb | Anti-sense | Chr. 16 - 76122613 - 76156086 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 30790 |
Gm30790 | 463.1kb | Sense | Chr. 16 - 76751793 - 76798855 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
