Assay Details
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082220.1 | - | 1 | 548 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuroendocrine secretory protein antisense | Nespas |
Gnas-as, Nespos |
-35.6kb | Anti-sense | Chr. 2 - 174281237 - 174295436 on NCBI Build GRCm38 |
Downstream |
negative elongation factor complex member C/D, Th1l |
Nelfcd |
2410003I03Rik, C77797, Th1, Th1l |
81.9kb | Sense | Chr. 2 - 174415804 - 174427503 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
