Assay Details
Gene Symbol: Gm15133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040749.1 | 4 - 5 | 318 | 78 | ||
GenBank mRNA | AK085986.1 | - | 4 - 5 | 319 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif-containing 12A | Trim12a |
2310043C01Rik, Trim12 |
-2.0kb | Anti-sense | Chr. 7 - 104299893 - 104315849 on NCBI Build GRCm38 |
Downstream | tripartite motif-containing 34B | Trim34b |
Gm15134, OTTMUSG00000019188, Trim34-2 |
10.6kb | Sense | Chr. 7 - 104329471 - 104336617 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
