Assay Details
Gene Symbol: Gm8179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046039.1 | 2 - 3 | 457 | 72 | ||
GenBank mRNA | AK143160.1 | - | 2 - 3 | 459 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26578 |
Gm26578 | -160.6kb | Sense | Chr. 8 - 32009913 - 32018405 on NCBI Build GRCm38 | |
Downstream | predicted gene 3985 | Gm3985 | 706.7kb | Anti-sense | Chr. 8 - 32888505 - 32950026 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61645 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752484 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204789 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386368 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75916 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316895 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9970 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61530 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033756 | AK143160.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06468861 | - | - |
Whole Mouse Genome G4122A | A_51_P185781 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06468861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06860480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07209576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06870628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06789157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06860461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07209586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06869735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07183839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06964843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762321 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07209574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06757313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07104445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07143418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07325449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07101589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07423804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25762287 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0800002873.mm.1 | TC0800000380.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800002873.mm.1 | NR_046039.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800002873.mm.1 | NR_046039.1 | Refseq |
