Assay Details
Gene Symbol: A830008E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154552.1 | - | 3 - 4 | 1785 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate-ammonia ligase (glutamine synthetase) | Glul |
GS, Glns |
-138.4kb | Anti-sense | Chr. 1 - 153899929 - 153909723 on NCBI Build GRCm38 |
Downstream | zinc finger protein 648 | Zfp648 |
ENSMUSG00000066797, Gm10178, Gm203 |
123.0kb | Anti-sense | Chr. 1 - 154201187 - 154212916 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
