Assay Details
Gene Symbol: Gm5833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040304.1 | 1 - 2 | 105 | 95 | ||
GenBank mRNA | AK154631.1 | - | 1 - 2 | 107 | 95 | |
AK171408.1 | - | 1 - 2 | 97 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NIMA (never in mitosis gene a)-related expressed kinase 7 | Nek7 |
2810460C19Rik, AU020186 |
-58.0kb | Sense | Chr. 1 - 138484106 - 138619779 on NCBI Build GRCm38 |
Downstream | LIM homeobox protein 9 | Lhx9 |
3110009O07Rik, BB104635, LH2B |
143.1kb | Sense | Chr. 1 - 138825186 - 138848649 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001516.rn.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
