Assay Details
Gene Symbol: AU022793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045719.1 | 2 - 3 | 358 | 149 | ||
GenBank mRNA | AK155489.1 | - | 2 - 3 | 361 | 149 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | low density lipoprotein-related protein 12 | Lrp12 |
AI848829, C820005L12Rik, LRP-12 |
-20.1kb | Anti-sense | Chr. 15 - 39870602 - 39944009 on NCBI Build GRCm38 |
Downstream |
predicted gene, 33251 |
Gm33251 | 78.2kb | Sense | Chr. 15 - 40045191 - 40128982 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232057 | BG068400.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232057 | AU022793.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2232057 | AK155489.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847872 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08343689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156836 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11985491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156839 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12331244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156832 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11985502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156888 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11985505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12156878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156765 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156900 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12156880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12259756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156785 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156811 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156801 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156773 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156840 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12139590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12112870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12156876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156908 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156909 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156844 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156895 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12050351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156777 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12016953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30156882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12382482 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1445977_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445977_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445977_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500002245.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002245.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002245.mm.1 | NR_045719.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002244.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002244.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002244.mm.1 | NR_045719.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002247.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002247.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002247.mm.1 | NR_045719.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002243.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002243.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002243.mm.1 | NR_045719.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002248.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002248.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002248.mm.1 | NR_045719.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002249.mm.1 | TC1500000311.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500002249.mm.1 | NR_045719.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500002249.mm.1 | NR_045719.1 | Refseq |
Mouse Expression Set 430 | 1445977_PM_at | - | - |
Mouse Expression Set 430 | 1445977_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445977_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445977_PM_at | - | - |
