Assay Details
Gene Symbol: F830002L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033558.1 | 1 - 2 | 137 | 86 | ||
GenBank mRNA | AK158018.1 | - | 1 - 2 | 137 | 86 | |
AK162643.1 | - | 1 - 2 | 139 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CD24a antigen | Cd24a |
Cd24, HSA, Ly-52, nectadrin |
-9.2kb | Sense | Chr. 10 - 43579169 - 43584265 on NCBI Build GRCm38 |
Downstream |
predicted gene, 34481 |
Gm34481 | 54.3kb | Sense | Chr. 10 - 43658852 - 43662397 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
