Assay Details
Gene Symbol: Akr1cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC028632.1 | - | 2 - 3 | 264 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
pleckstrin homology domain containing, family M, member 3 |
Plekhm3 |
9430067K14Rik, A230102O09Rik, AI449570, DAPR, Plekhm1l |
-70.9kb | Sense | Chr. 1 - 64789120 - 64956824 on NCBI Build GRCm38 |
Downstream |
crystallin, gamma E |
Cryge |
Cryg-2, Cryg-6, DGcry-2, Elo |
17.9kb | Sense | Chr. 1 - 65048555 - 65057116 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
