Assay Details
Gene Symbol: 2700063P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012482.1 | - | 1 | 145 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | paralemmin 2 | Palm2 |
53-H12, B130066I24, R75368, pEN70 |
Sense | Chr. 4 - 57568248 - 57717146 on NCBI Build GRCm38 | |
Downstream | A kinase (PRKA) anchor protein 2 | Akap2 |
AA959716, AI649048, Akap-2, Akap-kl, B230340M18Rik, Prka2 |
233.4kb | Sense | Chr. 4 - 57845248 - 57896984 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
