Assay Details
Gene Symbol: 3110001L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013958.1 | - | 1 | 453 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 40630 |
Gm40630 | -7.7kb | Anti-sense | Chr. 10 - 42041555 - 42081937 on NCBI Build GRCm38 | |
Downstream | forkhead box O3 | Foxo3 |
1110048B16Rik, 2010203A17Rik, C76856, FKHRL1, Fkhr2, Foxo3a |
91.6kb | Anti-sense | Chr. 10 - 42181845 - 42276746 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
