Assay Details
Gene Symbol: 4930421P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015174.1 | - | 1 - 2 | 472 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 8 |
Adamts8 |
METH-2, METH2 |
-66.1kb | Anti-sense | Chr. 9 - 30942563 - 30962858 on NCBI Build GRCm38 |
Downstream | zinc finger and BTB domain containing 44 | Zbtb44 |
6030404E16Rik, Btbd15, E230011F09 |
559.0 | Anti-sense | Chr. 9 - 31030394 - 31075885 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
