Assay Details
Gene Symbol: Tmem144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004282.1 | - | 1 | 453 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | relaxin/insulin-like family peptide receptor 1 | Rxfp1 |
Gm1018, Lgr7 |
-74.6kb | Sense | Chr. 3 - 79641600 - 79737951 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 198, member B |
Fam198b |
1110032E23Rik, 2210419I08Rik, AV011458, Ened |
71.0kb | Anti-sense | Chr. 3 - 79884475 - 79946278 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
