Assay Details
Gene Symbol: 1700120B22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006818.1 | - | 1 - 2 | 262 | 66 | |
AK018869.1 | - | 1 - 2 | 258 | 66 | ||
AK018967.1 | - | 1 - 2 | 262 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myopalladin | Mypn |
1110056A04Rik, 9830112J03, AI853556, mKIAA4170 |
-39.1kb | Sense | Chr. 10 - 63115795 - 63203952 on NCBI Build GRCm38 |
Downstream | sirtuin 1 | Sirt1 |
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha |
74.9kb | Sense | Chr. 10 - 63319005 - 63339061 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
