Assay Details
Gene Symbol: 8030431A06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020196.1 | - | 1 | 742 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
family with sequence similarity 170, member B |
Fam170b |
4922501K12Rik, AI449756 |
-27.9kb | Sense | Chr. 14 - 32833962 - 32836789 on NCBI Build GRCm38 |
Downstream | WD repeat and FYVE domain containing 4 | Wdfy4 | Gm18810 | 93.9kb | Anti-sense | Chr. 14 - 32959547 - 33193855 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
