Assay Details
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020581.1 | - | 1 | 344 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spastin | Spast |
Spg4, mKIAA1083 |
-4.5kb | Sense | Chr. 17 - 74337454 - 74391115 on NCBI Build GRCm38 |
Downstream |
NLR family, CARD domain containing 4 |
Nlrc4 |
9530011P19Rik, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, Card12, IPAF |
29.5kb | Anti-sense | Chr. 17 - 74425716 - 74459143 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
