Assay Details
Gene Symbol: Gm11959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019683.1 | - | 2 - 3 | 507 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leukemia inhibitory factor | Lif | -2.4kb | Sense | Chr. 11 - 4257568 - 4272514 on NCBI Build GRCm38 | |
Downstream | HORMA domain containing 2 | Hormad2 | 4930529M09Rik | 63.9kb | Anti-sense | Chr. 11 - 4345801 - 4442459 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
