Assay Details
Gene Symbol: Gm29904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132805.1 | - | 3 - 4 | 388 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | alpha-1-B glycoprotein | A1bg | C44 | -371.8kb | Anti-sense | Chr. 15 - 60917589 - 60921270 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA D030024E09 gene | D030024E09Rik | 404.7kb | Anti-sense | Chr. 15 - 61699284 - 61774451 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
