Assay Details
Gene Symbol: Gm16796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040367.1 | 1 - 2 | 1085 | 119 | ||
GenBank mRNA | AK138533.1 | - | 1 - 2 | 1086 | 119 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prefoldin 4 | Pfdn4 |
0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4 |
Anti-sense | Chr. 2 - 170496428 - 170519070 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930470P17 gene | 4930470P17Rik | 67.5kb | Sense | Chr. 2 - 170579418 - 170602017 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
